Learn More
n-Pentylcyclohexane, 98%
CAS: 4292-92-6 | C11H22 | 154.297 g/mol
Supplier: Thermo Scientific Chemicals B2132609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Pentylcyclohexane | |
| -58°C | |
| 202°C | |
| Mild | |
| 1.4445 | |
| 10 g | |
| n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl | |
| CCCCCC1CCCCC1 | |
| 154.297 | |
| 154.3 |
| 4292-92-6 | |
| 0.8 | |
| 63°C (145°F) | |
| C11H22 | |
| MFCD00019402 | |
| UN3082 | |
| HLTMUYBTNSVOFY-UHFFFAOYSA-N | |
| pentylcyclohexane | |
| 20284 | |
| 98% |
Chemical Identifiers
| 4292-92-6 | |
| 154.297 | |
| HLTMUYBTNSVOFY-UHFFFAOYSA-N | |
| 20284 | |
| CCCCCC1CCCCC1 |
| C11H22 | |
| MFCD00019402 | |
| n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl | |
| pentylcyclohexane |
Safety and Handling
P210-P280-P370+P378q-P501c
H227
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 224-296-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only