missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylcyclohexane, 98%

Catalog No. p-7025691
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2132604 2 g
AAB2132609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2132604 Supplier Thermo Scientific Chemicals Supplier No. B2132604
Only null left

CAS: 4292-92-6 | C11H22 | 154.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4292-92-6
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00019402
InChI Key HLTMUYBTNSVOFY-UHFFFAOYSA-N
Synonym n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
PubChem CID 20284
IUPAC Name pentylcyclohexane
SMILES CCCCCC1CCCCC1

Specifications

Melting Point -58°C
Density 0.8
Boiling Point 202°C
Flash Point 63°C (145°F)
Odor Mild
Refractive Index 1.4445
Quantity 2 g
UN Number UN3082
Formula Weight 154.3
Percent Purity 98%
Chemical Name or Material n-Pentylcyclohexane
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 224-296-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only