missing translation for 'onlineSavingsMsg'
Learn More

n-Pentyl 4-hydroxybenzoate, 98%

Catalog No. AAL0556314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0556314 25 g
AAL0556306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0556314 Supplier Thermo Scientific Chemicals Supplier No. L0556314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6521-29-5 | C12H16O3 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Pentyl 4-hydroxybenzoate, is used as an antimicrobial agent in cosmetic products.

Solubility
Insoluble in water. Slightly soluble in chloroform, methanol.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 6521-29-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00016479
InChI Key ZNSSPLQZSUWFJT-UHFFFAOYSA-N
PubChem CID 23019
IUPAC Name pentyl 4-hydroxybenzoate
SMILES CCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 36°C to 40°C
Boiling Point 197°C to 200°C (10 mmHg)
Quantity 25 g
Beilstein 2722438
Synonym amyl 4-hydroxybenzoate, pentyl p-hydroxybenzoate, pentylparaben, n-pentyl 4-hydroxybenzoate, benzoic acid, 4-hydroxy-, pentyl ester, p-hydroxybenzoic acid pentyl ester, pentyl paraben, unii-sjv2e4c7dm, p-oxybenzoesaurepentylester, amyl4-hydroxybenzoate
Solubility Information Insoluble in water. Slightly soluble in chloroform,methanol.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material n-Pentyl 4-hydroxybenzoate
EINECSNumber 229-408-9
RTECSNumber DH2555000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only