Learn More
n-Pentadecane, 99%
CAS: 629-62-9 | C15H32 | 212.42 g/mol
Supplier: Thermo Scientific Chemicals A1033618
Description
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| n-Pentadecane | |
| 9°C to 10°C | |
| 269°C to 270°C | |
| C15H32 | |
| MFCD00008990 | |
| 1698194 | |
| Immiscible with water. | |
| CCCCCCCCCCCCCCC | |
| 212.42 | |
| CHEBI:28897 | |
| 99% |
| 629-62-9 | |
| 0.769 | |
| 132°C (269°F) | |
| 1.432 | |
| 50 g | |
| n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n | |
| YCOZIPAWZNQLMR-UHFFFAOYSA-N | |
| pentadecane | |
| 12391 | |
| 212.42 |
Chemical Identifiers
| 629-62-9 | |
| 212.42 | |
| YCOZIPAWZNQLMR-UHFFFAOYSA-N | |
| 12391 | |
| pentadecane |
| C15H32 | |
| MFCD00008990 | |
| n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n | |
| CHEBI:28897 | |
| CCCCCCCCCCCCCCC |
Safety and Handling
GHS H Statement
H304
May be fatal if swallowed and enters airways.
P301+P310-P331-P501c
H304
EINECSNumber : 211-098-1
RTECSNumber : RZ1800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only