missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecane, 99%

Catalog No. p-7080698
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1033614 25 g
AAA1033618 50 g
AAA1033630 250 g
AAA1033636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1033614 Supplier Thermo Scientific Chemicals Supplier No. A1033614
Only null left

CAS: 629-62-9 | C15H32 | 212.42 g/mol

n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 629-62-9
Molecular Formula C15H32
Molecular Weight (g/mol) 212.42
MDL Number MFCD00008990
InChI Key YCOZIPAWZNQLMR-UHFFFAOYSA-N
Synonym n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
PubChem CID 12391
ChEBI CHEBI:28897
IUPAC Name pentadecane
SMILES CCCCCCCCCCCCCCC

Specifications

Melting Point 9°C to 10°C
Density 0.769
Boiling Point 269°C to 270°C
Flash Point 132°C (269°F)
Refractive Index 1.432
Quantity 25 g
Beilstein 1698194
Solubility Information Immiscible with water.
Formula Weight 212.42
Percent Purity 99%
Chemical Name or Material n-Pentadecane
Show More Show Less
Hazard Category H304
Hazard Statement GHS H Statement
H304
May be fatal if swallowed and enters airways.
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 211-098-1
RTECSNumber RZ1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only