missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)-L-phenylalanyl Chloride 97.0+%, TCI America™

Catalog No. T14445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14445G 5 g
T14441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14445G Supplier TCI America Supplier No. T14445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29739-88-6
Molecular Formula C16H16ClNO3S
Molecular Weight (g/mol) 337.818
MDL Number MFCD00191583
InChI Key KISOIDIHUAPEON-HNNXBMFYSA-N
Synonym Tosyl-L-phenylalanyl Chloride
PubChem CID 7019169
IUPAC Name (2S)-2-[(4-methylphenyl)sulfonylamino]-3-phenylpropanoyl chloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 337.82
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanyl Chloride [Optical Resolving Reagent for Alcohols]