accessibility menu, dialog, popup

UserName

CH3SOONHOCl

CAS RN 29739-88-6

IUPAC Name: 2-(4-methylbenzenesulfonamido)-3-phenylpropanoyl chloride
Synonyms: 2-(4-methylbenzenesulfonamido)-3-phenylpropanoyl chloride
Molecular Weight (g/mol): 337.82
Molecular Formula: C16H16ClNO3S
InChi Key: KISOIDIHUAPEON-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(CC1=CC=CC=C1)C(Cl)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results