missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(p-Toluenesulfonyl)-L-phenylalanyl Chloride 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 132°C |
| Color | White-Yellow |
| Quantity | 1 g |
| UN Number | 3261 |
| Formula Weight | 337.82 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(p-Toluenesulfonyl)-L-phenylalanyl Chloride [Optical Resolving Reagent for Alcohols] |
Safety and Handling
| TSCA | No |