missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)-L-phenylalanine 98.0+%, TCI America™

Catalog No. p-7136779
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T144325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T144325G Supplier TCI America Supplier No. T144325G

Chemical Identifiers

CAS 13505-32-3
Molecular Formula C16H17NO4S
Molecular Weight (g/mol) 319.375
MDL Number MFCD00037251
InChI Key CGRCVIZBNRUWLY-HNNXBMFYSA-N
Synonym Tosyl-L-phenylalanine
PubChem CID 99327
IUPAC Name (2S)-2-[(4-methylphenyl)sulfonylamino]-3-phenylpropanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 25 g
Formula Weight 319.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanine