accessibility menu, dialog, popup

UserName

CAS RN 13505-32-3

IUPAC Name: 2-(4-methylbenzenesulfonamido)-3-phenylpropanoic acid
Synonyms: 2-(4-methylbenzenesulfonamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 319.38
Molecular Formula: C16H17NO4S
InChi Key: CGRCVIZBNRUWLY-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results