missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(p-Toluenesulfonyl)glycine 98.0+%, TCI America™
$57.79 - $164.03
Chemical Identifiers
| CAS | 1080-44-0 |
|---|---|
| Molecular Formula | C9H11NO4S |
| Molecular Weight (g/mol) | 229.25 |
| MDL Number | MFCD00045898 |
| InChI Key | VDKFCCZUCXYILI-UHFFFAOYSA-N |
| Synonym | n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid |
| PubChem CID | 70653 |
| IUPAC Name | 2-[(4-methylphenyl)sulfonylamino]acetic acid |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O |
Chemical Identifiers
| 1080-44-0 | |
| 229.25 | |
| VDKFCCZUCXYILI-UHFFFAOYSA-N | |
| 70653 | |
| CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O |
| C9H11NO4S | |
| MFCD00045898 | |
| n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid | |
| 2-[(4-methylphenyl)sulfonylamino]acetic acid |
Specifications
| 1080-44-0 | |
| C9H11NO4S | |
| 5 g | |
| VDKFCCZUCXYILI-UHFFFAOYSA-N | |
| 2-[(4-methylphenyl)sulfonylamino]acetic acid | |
| 70653 | |
| ≥98.0% (HPLC,T) | |
| N-(p-Toluenesulfonyl)glycine |
| 149°C | |
| MFCD00045898 | |
| n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid | |
| CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O | |
| 229.25 | |
| 229.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No