missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)glycine 98.0+%, TCI America™

Catalog No. T28035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28035G 5 g
T280325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28035G Supplier TCI America Supplier No. T28035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1080-44-0
Molecular Formula C9H11NO4S
Molecular Weight (g/mol) 229.25
MDL Number MFCD00045898
InChI Key VDKFCCZUCXYILI-UHFFFAOYSA-N
Synonym n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid
PubChem CID 70653
IUPAC Name 2-[(4-methylphenyl)sulfonylamino]acetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O

Specifications

Melting Point 149°C
Quantity 5 g
Formula Weight 229.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluenesulfonyl)glycine