missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrichlorosilane, 97%

Catalog No. p-7027191
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAB2375314 25 g
AAB2375322 100 g
AAB2375336 500 g
3 options

Catalog No. AAB2375314

Supplier: Thermo Scientific Chemicals B2375314

Only null left

CAS: 5283-66-9 | C8H17Cl3Si | 247.659 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5283-66-9
Molecular Formula C8H17Cl3Si
Molecular Weight (g/mol) 247.659
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 225°C to 227°C
Quantity 25 g
UN Number UN1801
Beilstein 1744230
Flash Point 96°C (204°F)
Formula Weight 247.67
Percent Purity 97%
Odor Irritating
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 1.073
Chemical Name or Material n-Octyltrichlorosilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTYLTRICHLOROSILANE
EINECSNumber 226-112-1
RTECSNumber VV4700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only