missing translation for 'onlineSavingsMsg'
Learn More

n-Octylbenzene 98.0+%, TCI America™

Catalog No. p-7134153
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
O004625ML 25 mL
O0046100ML 100 mL
O0046500ML 500 mL
3 options

Catalog No. O004625ML

Supplier: TCI America O004625ML

Only null left

Chemical Identifiers

CAS 2189-60-8
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00009564
InChI Key CDKDZKXSXLNROY-UHFFFAOYSA-N
Synonym n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
PubChem CID 16607
IUPAC Name octylbenzene
SMILES CCCCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 mL
Formula Weight 190.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octylbenzene
EINECSNumber (3)-0021
TSCA Yes