accessibility menu, dialog, popup

UserName

CAS RN 2189-60-8

IUPAC Name: octylbenzene
Synonyms: octylbenzene
Molecular Weight (g/mol): 190.33
Molecular Formula: C14H22
InChi Key: CDKDZKXSXLNROY-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)