missing translation for 'onlineSavingsMsg'
Learn More

n-Octylbenzene 98.0+%, TCI America™

Catalog No. O004625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O004625ML 25 mL
O0046100ML 100 mL
O0046500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O004625ML Supplier TCI America Supplier No. O004625ML

Chemical Identifiers

CAS 2189-60-8
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00009564
InChI Key CDKDZKXSXLNROY-UHFFFAOYSA-N
Synonym n-octylbenzene, 1-phenyloctane, benzene, octyl, phenyloctane, octane, 1-phenyl, octyl-benzene, 1-octylbenzene, octyl benzene, 4-octylbenzene, acmc-1cplg
PubChem CID 16607
IUPAC Name octylbenzene
SMILES CCCCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 264°C
Quantity 25 mL
Formula Weight 190.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octylbenzene
EINECSNumber (3)-0021
TSCA Yes