missing translation for 'onlineSavingsMsg'
Learn More

n-Octylamine 98.0+%, TCI America™

Catalog No. O004525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O004525ML 25 mL
O0045500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O004525ML Supplier TCI America Supplier No. O004525ML

Chemical Identifiers

CAS 111-86-4
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008247
InChI Key IOQPZZOEVPZRBK-UHFFFAOYSA-N
Synonym octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
PubChem CID 8143
ChEBI CHEBI:7728
IUPAC Name octan-1-amine
SMILES CCCCCCCCN

Specifications

Color Yellow
Boiling Point 176°C
Quantity 25 mL
UN Number 2735
Formula Weight 129.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octylamine
EINECSNumber (2)-0133
RTECSNumber RG8050000
TSCA Yes