missing translation for 'onlineSavingsMsg'
Learn More

n-Octylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134359
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04841G 1 g
O04845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04841G Supplier TCI America Supplier No. O04841G

Chemical Identifiers

CAS 142-95-0
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.71
MDL Number MFCD00042019
InChI Key PHFDTSRDEZEOHG-UHFFFAOYSA-N
Synonym octylammonium chloride, octylamine hydrochloride, 1-octanamine, hydrochloride, octylamine, hydrochloride, octylamine-monohydrochloride, 1-octanamine, hydrochloride 1:1, octan-1-aminium chloride, octylamine, chloride, n-octylammonium chloride, n-octylamine hydrochloride
PubChem CID 67346
IUPAC Name hydrogen octan-1-amine chloride
SMILES [H+].[Cl-].CCCCCCCCN

Specifications

Melting Point 198°C
Color White
Quantity 1 g
Formula Weight 165.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octylamine Hydrochloride