accessibility menu, dialog, popup

UserName

CAS RN 142-95-0

IUPAC Name: hydrogen octan-1-amine chloride
Synonyms: hydrogen octylamine chloride
Molecular Weight (g/mol): 165.71
Molecular Formula: C8H20ClN
InChi Key: PHFDTSRDEZEOHG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results