missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl Sulfide 97.0+%, TCI America™
$33.56 - $169.63
Chemical Identifiers
| CAS | 2690-08-6 |
|---|---|
| Molecular Formula | C16H34S |
| Molecular Weight (g/mol) | 258.51 |
| MDL Number | MFCD00009566 |
| InChI Key | LOXRGHGHQYWXJK-UHFFFAOYSA-N |
| Synonym | dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane |
| PubChem CID | 75901 |
| IUPAC Name | 1-(octylsulfanyl)octane |
| SMILES | CCCCCCCCSCCCCCCCC |
Chemical Identifiers
| 2690-08-6 | |
| 258.51 | |
| LOXRGHGHQYWXJK-UHFFFAOYSA-N | |
| 75901 | |
| CCCCCCCCSCCCCCCCC |
| C16H34S | |
| MFCD00009566 | |
| dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane | |
| 1-(octylsulfanyl)octane |
Specifications
| 2690-08-6 | |
| 309°C | |
| MFCD00009566 | |
| dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane | |
| CCCCCCCCSCCCCCCCC | |
| 258.51 | |
| 258.51 | |
| Liquid |
| Yellow | |
| C16H34S | |
| 10 g | |
| LOXRGHGHQYWXJK-UHFFFAOYSA-N | |
| 1-(octylsulfanyl)octane | |
| 75901 | |
| ≥97.0% (GC) | |
| n-Octyl Sulfide |
Safety and Handling
EINECSNumber : (2)-0472
TSCA : Yes