missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl Gallate 98.0+%, TCI America™
$113.71 - $438.05
Chemical Identifiers
| CAS | 1034-01-1 |
|---|---|
| Molecular Formula | C15H22O5 |
| Molecular Weight (g/mol) | 282.336 |
| MDL Number | MFCD00002197 |
| InChI Key | NRPKURNSADTHLJ-UHFFFAOYSA-N |
| Synonym | octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove |
| PubChem CID | 61253 |
| ChEBI | CHEBI:83631 |
| IUPAC Name | octyl 3,4,5-trihydroxybenzoate |
| SMILES | CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O |
Chemical Identifiers
| 1034-01-1 | |
| 282.336 | |
| NRPKURNSADTHLJ-UHFFFAOYSA-N | |
| 61253 | |
| octyl 3,4,5-trihydroxybenzoate |
| C15H22O5 | |
| MFCD00002197 | |
| octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove | |
| CHEBI:83631 | |
| CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O |
Specifications
| 1034-01-1 | |
| C15H22O5 | |
| 25 g | |
| NRPKURNSADTHLJ-UHFFFAOYSA-N | |
| octyl 3,4,5-trihydroxybenzoate | |
| 61253 | |
| 282.34 | |
| Crystalline Powder |
| 100°C | |
| MFCD00002197 | |
| octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove | |
| CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O | |
| 282.336 | |
| CHEBI:83631 | |
| ≥98.0% (T) | |
| n-Octyl Gallate |
Safety and Handling
EINECSNumber : (3)-3071
RTECSNumber : LW8225000
TSCA : Yes