missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl gallate, 98+%

CAS: 1034-01-1 | C15H22O5 | 282.336 g/mol

$109.45 - $282.45

Chemical Identifiers

CAS 1034-01-1
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00002197
InChI Key NRPKURNSADTHLJ-UHFFFAOYSA-N
Synonym octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
PubChem CID 61253
ChEBI CHEBI:83631
IUPAC Name octyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0608114
View Documents
Thermo Scientific Chemicals
L0608114
25 g
Each for $109.45
Only null left
 
AAL0608122
View Documents
Thermo Scientific Chemicals
L0608122
100 g
Each for $282.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1034-01-1
282.336
NRPKURNSADTHLJ-UHFFFAOYSA-N
61253
octyl 3,4,5-trihydroxybenzoate
C15H22O5
MFCD00002197
octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
CHEBI:83631
CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

1034-01-1
C15H22O5
25 g
octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O
282.336
CHEBI:83631
≥98%
96°C to 102°C
MFCD00002197
2132305
NRPKURNSADTHLJ-UHFFFAOYSA-N
octyl 3,4,5-trihydroxybenzoate
61253
282.34
n-Octyl gallate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 213-853-0

RTECSNumber : LW8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only