missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Ether 96.0+%, TCI America™

Supplier:  TCI America O004725ML

 View more versions of this product

Catalog No. O004725ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

n-Octyl Ether
-7°C
287°C
MFCD00009563
Di-n-octyl Ether
CCCCCCCCOCCCCCCCC
242.447
242.45
Liquid
629-82-3
Yellow
C16H34O
25 mL
NKJOXAZJBOMXID-UHFFFAOYSA-N
1-octoxyoctane
12399
≥96.0% (GC)

Chemical Identifiers

629-82-3
242.447
NKJOXAZJBOMXID-UHFFFAOYSA-N
12399
CCCCCCCCOCCCCCCCC
C16H34O
MFCD00009563
Di-n-octyl Ether
1-octoxyoctane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3074

RTECSNumber : RH8800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions