missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl Ether 96.0+%, TCI America™
Supplier: TCI America O0047100ML
Specifications
| n-Octyl Ether | |
| -7°C | |
| 287°C | |
| MFCD00009563 | |
| Di-n-octyl Ether | |
| CCCCCCCCOCCCCCCCC | |
| 242.447 | |
| 242.45 | |
| Liquid |
| 629-82-3 | |
| Yellow | |
| C16H34O | |
| 100 mL | |
| NKJOXAZJBOMXID-UHFFFAOYSA-N | |
| 1-octoxyoctane | |
| 12399 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 629-82-3 | |
| 242.447 | |
| NKJOXAZJBOMXID-UHFFFAOYSA-N | |
| 12399 | |
| CCCCCCCCOCCCCCCCC |
| C16H34O | |
| MFCD00009563 | |
| Di-n-octyl Ether | |
| 1-octoxyoctane |
Safety and Handling
EINECSNumber : (2)-3074
RTECSNumber : RH8800000
TSCA : Yes