missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Octyl-β-D-glucopyranoside, ULTROL™ Grade, Calbiochem™,

Catalog No. 4944601GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4944601GM 1 g Glass Bottle
4944605GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4944601GM Supplier MilliporeSigma™ Supplier No. 4944601GM

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym OG, n-Octylglucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥99% (TLC)
Grade ULTROL