missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ n-Octyl-β-D-glucopyranoside, Calbiochem™,
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Packaging | Glass Bottle |
| Percent Purity | ≥98% (HPLC) |
| Color | White |
| Quantity | 25 g |
| Packaging | Glass Bottle |
| Percent Purity | ≥98% (HPLC) |