missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-β-D-Glucopyranoside, MP Biomedicals™

Catalog No. p-4404621
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15005401 1 g
ICN15005405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15005401 Supplier MP Biomedicals Inc Supplier No. 0215005401

n-Octyl-β-D-Glucopyranoside >98%, white powder, non-ionic detergent for membrane protein.

  • Non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids.
  • Used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers.
  • Used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym n-Octyl-b-D-glucoside, OGP
PubChem CID 62852
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Physical Form Powder
Melting Point 89°C to 100°C (Literature)
Quantity 1 g
Assay Percent Range ≥98%
Specific Rotation -31 ± 3° (c=5, water)
Beilstein 84118
Solubility Information Soluble in water (10mg/mL-clear colorless solution).
Chemical Name or Material n-Octyl-β-D-glucopyranoside, Molecular Biology
Grade 98%
EINECSNumber 249-887-8
storageNote1 20°C
Recommended Storage Store at -0°C.