missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl butyrate, 97%

CAS: 110-39-4 | C12H24O2 | 200.322 g/mol

Chemical Identifiers

CAS 110-39-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00048939
InChI Key PWLNAUNEAKQYLH-UHFFFAOYSA-N
Synonym octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
PubChem CID 61030
IUPAC Name octyl butanoate
SMILES CCCCCCCCOC(=O)CCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAB2497818
View Documents Promotion Details
N/A 50 g N/A
Only null left
 
AAB2497830
View Documents Promotion Details
N/A 250 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-39-4
200.322
PWLNAUNEAKQYLH-UHFFFAOYSA-N
61030
CCCCCCCCOC(=O)CCC
C12H24O2
MFCD00048939
octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
octyl butanoate

Specifications

110-39-4
MFCD00048939
PWLNAUNEAKQYLH-UHFFFAOYSA-N
octyl butanoate
61030
97%
C12H24O2
octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
CCCCCCCCOC(=O)CCC
200.322
172.27
n-Octyl butyrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-762-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only