missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl butyrate, 97%

Catalog No. AAB2497818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2497818 50 g
AAB2497830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2497818 Supplier Thermo Scientific Chemicals Supplier No. B2497818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-39-4 | C12H24O2 | 200.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-39-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00048939
InChI Key PWLNAUNEAKQYLH-UHFFFAOYSA-N
Synonym octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
PubChem CID 61030
IUPAC Name octyl butanoate
SMILES CCCCCCCCOC(=O)CCC

Specifications

Quantity 50 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material n-Octyl butyrate
EINECSNumber 203-762-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only