missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-thioglucopyranoside, 98+%

Non-ionic detergent used for the solubilization of membrane proteins | CAS: 85618-21-9 | C14H28O5S | 308.433 g/mol

Supplier:  Thermo Scientific Chemicals J6102806

 View more versions of this product

Catalog No. AAJ6102806


Only null left

Description

Description

n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Solubility
Soluble in ethanol, water or methanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

n-Octyl-beta-D-thioglucopyranoside
128°C
C14H28O5S
5 g
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
CGVLVOOFCGWBCS-DGTMBMJNSA-N
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
53720324
≥98%
85618-21-9
White
MFCD00012189
4182783
Soluble in ethanol, water or methanol
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
308.433
308.44
Crystalline Powder

Chemical Identifiers

85618-21-9
308.433
CGVLVOOFCGWBCS-DGTMBMJNSA-N
53720324
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
C14H28O5S
MFCD00012189
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only