missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-thioglucopyranoside, 98+%

Non-ionic detergent used for the solubilization of membrane proteins | CAS: 85618-21-9 | C14H28O5S | 308.433 g/mol

$407.90 - $1,136.58

Chemical Identifiers

CAS 85618-21-9
Molecular Formula C14H28O5S
Molecular Weight (g/mol) 308.433
MDL Number MFCD00012189
InChI Key CGVLVOOFCGWBCS-DGTMBMJNSA-N
Synonym octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
PubChem CID 53720324
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
SMILES CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6102888
View Documents Promotion Details
N/A 2.5 g
Each for $407.90
Only null left
 
AAJ6102806
View Documents Promotion Details
N/A 5 g
Each for $552.52
Only null left
 
AAJ6102809
View Documents Promotion Details
N/A 10 g
Each for $1,136.58
Only null left
 
Description

Description

n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Solubility
Soluble in ethanol, water or methanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

85618-21-9
308.433
CGVLVOOFCGWBCS-DGTMBMJNSA-N
53720324
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
C14H28O5S
MFCD00012189
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol

Specifications

85618-21-9
White
MFCD00012189
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
CGVLVOOFCGWBCS-DGTMBMJNSA-N
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
53720324
≥98%
n-Octyl-beta-D-thioglucopyranoside
128°C
C14H28O5S
4182783
Soluble in ethanol, water or methanol
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
308.433
308.44
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only