missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 57°C |
| Quantity | 500 mL |
| Formula Weight | 184.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | n-Octyl Acrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-0990 |
| TSCA | Yes |