missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. O047825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O047825ML 25 mL
O0478500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O047825ML Supplier TCI America Supplier No. O047825ML
Only null left

Chemical Identifiers

CAS 2499-59-4
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00048935
InChI Key ANISOHQJBAQUQP-UHFFFAOYSA-N
Synonym Acrylic Acid n-Octyl Ester
PubChem CID 17258
IUPAC Name octyl prop-2-enoate
SMILES CCCCCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 57°C
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0990
TSCA Yes