missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Acetate 98.0+%, TCI America™

Supplier:  TCI America A0042500ML

 View more versions of this product

Catalog No. A0042500ML


Only null left

Description

Description

Specifications

Specifications

n-Octyl Acetate
Colorless
C10H20O2
500 mL
YLYBTZIQSIBWLI-UHFFFAOYSA-N
octyl acetate
8164
172.27
Liquid
112-14-1
210°C
MFCD00009562
n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
CCCCCCCCOC(C)=O
172.27
CHEBI:87495
≥98.0% (GC)

Chemical Identifiers

112-14-1
172.27
YLYBTZIQSIBWLI-UHFFFAOYSA-N
8164
octyl acetate
C10H20O2
MFCD00009562
n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
CHEBI:87495
CCCCCCCCOC(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0735

RTECSNumber : AJ1400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions