missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Acetate 98.0+%, TCI America™

Catalog No. p-7116678
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A004225ML 25 mL
A0042500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A004225ML Supplier TCI America Supplier No. A004225ML
Only null left

Chemical Identifiers

CAS 112-14-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009562
InChI Key YLYBTZIQSIBWLI-UHFFFAOYSA-N
Synonym n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
PubChem CID 8164
ChEBI CHEBI:87495
IUPAC Name octyl acetate
SMILES CCCCCCCCOC(C)=O

Specifications

Color Colorless
Boiling Point 210°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Acetate
EINECSNumber (2)-0735
RTECSNumber AJ1400000
TSCA Yes