missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Octanoyl-N-methyl-D-glucamine

Catalog No. p-7042733
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6357403 1 g
AAJ6357406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6357403 Supplier Thermo Scientific Chemicals Supplier No. J6357403
Only null left

Non-ionic detergent for solubilizing membrane proteins | CAS: 85316-98-9 | C15H31NO6 | 321.414 g/mol

N-Octanoyl-N-methylglucamine has been used in a study to assess disordered the N-terminus of PKA. It has also been used in a study to investigate the usefulness of sugar surfactants as solubilizing agents in parenteral formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Octanoyl-N-methylglucamine has been used in a study to assess disordered the N-terminus of PKA. It has also been used in a study to investigate the usefulness of sugar surfactants as solubilizing agents in parenteral formulations.

Solubility
Soluble in water (10%).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 85316-98-9
Molecular Formula C15H31NO6
Molecular Weight (g/mol) 321.414
MDL Number MFCD00134152
InChI Key SBWGZAXBCCNRTM-CTHBEMJXSA-N
Synonym mega-8, n-octanoyl-n-methylglucamine, mega 8, mega-8 solution, n-methyl-n-octanoyl-d-glucamine, n-d-glucityl-n-methyloctanamide, mega-8 500 mm solution, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl octanamide, octanoyl-n-methylglucamide
PubChem CID 160091
IUPAC Name N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
SMILES CCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O

Specifications

Melting Point 80°C to 90°C
Color White
Quantity 1 g
Beilstein 5816747
Solubility Information Soluble in water (10%).
Optical Rotation −18° (c=2 in Water)
Formula Weight 321.41
Physical Form Powder
Chemical Name or Material N-Octanoyl-N-methyl-D-glucamine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only