missing translation for 'onlineSavingsMsg'
Learn More

n-Octanoyl Chloride 99.0+%, TCI America™

Catalog No. p-7134147
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003925ML 25 mL
O0039100ML 100 mL
O0039500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O003925ML Supplier TCI America Supplier No. O003925ML
Only null left

Chemical Identifiers

CAS 111-64-8
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.657
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 195°C
Quantity 25 mL
UN Number 3265
Formula Weight 162.66
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octanoyl Chloride
EINECSNumber (2)-0632
RTECSNumber RH1570000
TSCA Yes