missing translation for 'onlineSavingsMsg'
Learn More

n-Octanophenone 98.0+%, TCI America™

Catalog No. p-7134210
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
O014925ML 25 mL
O0149100ML 100 mL
O0149500ML 500 mL
3 options

Catalog No. O014925ML

Supplier: TCI America O014925ML

Only null left

Chemical Identifiers

CAS 1674-37-9
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00003554
InChI Key UDEVCZRUNOLVLU-UHFFFAOYSA-N
Synonym octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
PubChem CID 74291
IUPAC Name 1-phenyloctan-1-one
SMILES CCCCCCCC(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 290°C
Quantity 25 mL
Formula Weight 204.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material n-Octanophenone