missing translation for 'onlineSavingsMsg'
Learn More

n-Octanophenone 98.0+%, TCI America™

Catalog No. O014925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O014925ML 25 mL
O0149100ML 100 mL
O0149500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O014925ML Supplier TCI America Supplier No. O014925ML

Chemical Identifiers

CAS 1674-37-9
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00003554
InChI Key UDEVCZRUNOLVLU-UHFFFAOYSA-N
Synonym octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
PubChem CID 74291
IUPAC Name 1-phenyloctan-1-one
SMILES CCCCCCCC(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 290°C
Quantity 25 mL
Formula Weight 204.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material n-Octanophenone