missing translation for 'onlineSavingsMsg'
Learn More

n-Octanoic Anhydride 95.0+%, TCI America™

Catalog No. O003525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003525ML 25 mL
O0035500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003525ML Supplier TCI America Supplier No. O003525ML

Chemical Identifiers

CAS 623-66-5
Molecular Formula C16H30O3
Molecular Weight (g/mol) 270.413
MDL Number MFCD00056161
InChI Key RAFYDKXYXRZODZ-UHFFFAOYSA-N
PubChem CID 69340
IUPAC Name octanoyl octanoate
SMILES CCCCCCCC(=O)OC(=O)CCCCCCC

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 180°C
Quantity 25 mL
UN Number 3265
Formula Weight 270.41
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octanoic Anhydride