accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 623-66-5

IUPAC Name: octanoyl octanoate
Synonyms: octanoic acid, anhydride
Molecular Weight (g/mol): 270.41
Molecular Formula: C16H30O3
InChi Key: RAFYDKXYXRZODZ-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC(=O)CCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results