missing translation for 'onlineSavingsMsg'
Learn More

n-Octanoic Acid 98.0+%, TCI America™

Catalog No. O002725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002725ML 25 mL
O0027500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O002725ML Supplier TCI America Supplier No. O002725ML

Chemical Identifiers

CAS 124-07-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00004429
InChI Key WWZKQHOCKIZLMA-UHFFFAOYSA-N
Synonym caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
PubChem CID 379
ChEBI CHEBI:28837
IUPAC Name octanoic acid
SMILES CCCCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 238°C
Quantity 25 mL
UN Number 3265
Formula Weight 144.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octanoic Acid
EINECSNumber (2)-0608
RTECSNumber RH0175000
TSCA Yes