missing translation for 'onlineSavingsMsg'
Learn More

n-Octane 97.0+%, TCI America™

Catalog No. p-7134133
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
O002225ML 25 mL
O0022100ML 100 mL
O0022500ML 500 mL
3 options

Catalog No. O002225ML

Supplier: TCI America O002225ML

Only null left

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Melting Point -57°C
Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octane
EINECSNumber (2)-0008
RTECSNumber RG8400000
TSCA Yes