missing translation for 'onlineSavingsMsg'
Learn More

n-Octane 97.0+%, TCI America™

Catalog No. O002225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002225ML 25 mL
O0022100ML 100 mL
O0022500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O002225ML Supplier TCI America Supplier No. O002225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Melting Point -57°C
Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octane
EINECSNumber (2)-0008
RTECSNumber RG8400000
TSCA Yes