missing translation for 'onlineSavingsMsg'
Learn More

n-Octane 99.5+%, TCI America™

Catalog No. s02805ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02805ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02805ML Supplier TCI America Supplier No. S02805ML
Only null left

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Physical Form Liquid
UN Number 1262
Color Colorless
Melting Point -57°C
Boiling Point 125°C
Quantity 5 mL
Formula Weight 114.23
Percent Purity ≥99.5% (GC)
Chemical Name or Material n-Octane [Standard Material for GC]
EINECSNumber (2)-0008
RTECSNumber RG8400000
TSCA Yes