missing translation for 'onlineSavingsMsg'
Learn More

n-Octane 98.5+%, TCI America™

Supplier:  TCI America O0151500ML

Catalog No. O0151500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

n-Octane [for HPLC Solvent]
-57°C
125°C
MFCD00009556
1262
TVMXDCGIABBOFY-UHFFFAOYSA-N
octane
356
114.23
Liquid
111-65-9
Colorless
C8H18
500 mL
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
CCCCCCCC
114.232
CHEBI:17590
≥98.5% (GC)

Chemical Identifiers

111-65-9
114.232
TVMXDCGIABBOFY-UHFFFAOYSA-N
356
octane
C8H18
MFCD00009556
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
CHEBI:17590
CCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0008

RTECSNumber : RG8400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions