missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, EMD-Millipore

Catalog No. m1097160005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1097160005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1097160005 Supplier Technidata Supplier No. 1.09716.0005
Only null left

Hydrocarbons C8

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 0.8 to 6.5 %(V)
Ignition Point 210°C
Vapor Pressure 14 hPa (20°C)
Density Density: 0.70g/cm3 (20°)
Assay Percent Range ≥99.0%
Linear Formula CH3(CH2)6CH3
Quantity 5 mL
Formula Weight 114.23
Chemical Name or Material n-Octane
Recommended Storage Store without restrictions