missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, 98+%

CAS: 111-65-9 | C8H18 | 114.232 g/mol

Supplier:  Thermo Scientific Chemicals A13181AE

 View more versions of this product

Catalog No. AAA13181AE


Only null left
Explore available promotions

Description

Description

N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

Solubility
Miscible with ethyl ether, ethanol, acetone, benzene, petroleum ether and gasoline. Slightly Miscible with alcohol. Immiscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents and acids
Specifications

Specifications

n-Octane
-57°C
125°C to 126°C
≥98%
C8H18
MFCD00009556
UN1262
14,6749
Miscible with ethyl ether,ethanol,acetone,benzene,petroleum ether and gasoline; Slightly Miscible with alcohol; Immiscible with water.
CCCCCCCC
114.232
CHEBI:17590
≥98%
111-65-9
0.703
15°C (59°F)
Nearly Odorless
1.398
100 mL
1696875
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
TVMXDCGIABBOFY-UHFFFAOYSA-N
octane
356
114.23
Liquid

Chemical Identifiers

111-65-9
114.232
TVMXDCGIABBOFY-UHFFFAOYSA-N
356
octane
C8H18
MFCD00009556
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
CHEBI:17590
CCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES

EINECSNumber : 203-892-1

RTECSNumber : RG8400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only