missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, 97%, pure

CAS: 111-65-9 | C8H18 | 114.23 g/mol

Supplier:  Thermo Scientific Chemicals 325950025

 View more versions of this product

Catalog No. AC325950025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Octane
111-65-9
100.0
Colorless
125.0°C to 127.0°C
Authentic
Glass Bottle
1.3965 to 1.3985
MFCD00009556
01,159
15,6838
Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline
CCCCCCCC
114.23
0.55 mPa.s (20°C)
114.23
Pure
97%
96.0
-57.0°C
0.7080g/mL
13°C
96% min. (GC)
C8H18
CH3(CH2)6CH3
2.5 L
0.708
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
TVMXDCGIABBOFY-UHFFFAOYSA-N
octane
356
CHEBI:17590
97%
Liquid

Chemical Identifiers

111-65-9
114.23
TVMXDCGIABBOFY-UHFFFAOYSA-N
356
octane
C8H18
MFCD00009556
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
CHEBI:17590
CCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Danger

EINECSNumber : 203-892-1

RTECSNumber : RG8400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only