missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyltrimethylammonium chloride, 95%

Catalog No. p-7040793
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6066504 2 g
AAH6066509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6066504 Supplier Thermo Scientific Chemicals Supplier No. H6066504

CAS: 112-03-8 | C21H46ClN | 348.06 g/mol

Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

Solubility
Soluble in water 1.759 mg/L @ 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 112-03-8
Molecular Formula C21H46ClN
Molecular Weight (g/mol) 348.06
MDL Number MFCD00050188
InChI Key VBIIFPGSPJYLRR-UHFFFAOYSA-M
Synonym n,n,n-trimethyloctadecan-1-aminium chloride, stac, octadecyltrimethylammonium chloride, cation ab, stearyltrimethylammonium chloride, steartrimonium chloride, trimethyloctadecylammonium chloride, 1-octadecanaminium, n,n,n-trimethyl-, chloride, nissan cation ab, aliquat 7
PubChem CID 8155
ChEBI CHEBI:81710
IUPAC Name trimethyl(octadecyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Linear Formula CH3(CH2)17N(Cl)(CH3)3
Quantity 2 g
Beilstein 3917847
Solubility Information Soluble in water 1.759mg/L @ 25°C.
Formula Weight 348.05
Percent Purity 95%
Chemical Name or Material n-Octadecyltrimethylammonium chloride
Hazard Category H302-H311-H314-H335
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-929-1
RTECSNumber BT1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only